Electroporation cuvettes
Supplier: Thermo Fisher Scientific
5510-11 5520 5540-11
732-2267EA 250 GBP
732-2267 732-2268 732-2269
Electroporation cuvettes
Cuvettes Electroporation Cuvettes
Compatible with all common electroporators.
  • Precise gap tolerances ensure reproducible field strength delivery and high transformation efficiency
  • Consistent chamber shape made from seamless plastic moulding eliminates leaks and keeps the aluminium plates parallel for uniform sample treatment and safety
  • Colour-coded, frosted caps allow easy labelling and recognition
  • Caps are snug fitting to ensure sterility is maintained
Ordering information: Available in 3 gap sizes: 1 mm (white cap) delivers pulses with the highest field strength and is ideal forMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603)