J.T.Baker® BAKERBOND spe™ Sephadex® G-25, SPE Columns
Supplier: Avantor
7310-07EA 248 GBP
7310-07
J.T.Baker® BAKERBOND spe™ Sephadex® G-25, SPE Columns
Chromatography Columns
BAKERBOND spe™ columns are disposable PP columns pre-packed with high capacity sorbents contained between two 20 µm PE frits.
  • Silica based
  • Extraction of compounds from sample solution volumes ranging from a few hundred µl up to 1 L
  • Processing by vacuum, positive pressure or centrifugation
General applications: Desalting, removal of small molecular weight compounds.
These SPE columns can be used in size exclusion mode.
Sephadex® G-25 is a gel filtration media of cross-linked dextran. It is a hydrophilic packaging material, which swells in waterMore Product Information