SOLA™ SPE Plates, 96-Well
Supplier: Thermo Fisher Scientific
60309-003 60309-002 60309-001 60309-004 60309-005
554-1988EA 266 GBP
554-1988 554-1987 554-1986 730-1419 730-1420
SOLA™ SPE Plates, 96-Well
Microplates Solid Phase Extraction Plates (SPE)
SOLA™ fritless 96-well SPE plates provide greater reproducibility with cleaner, more consistent extracts. SOLA™ products provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
  • Higher levels of reproducibility
  • Higher levels of extract cleanliness
  • Reduced solvent requirements
  • Increased sensitivity
Order Now
SPECIFICATIONS
Environmentally Preferable