Chromeazurol S (Mordant blue 29) Indicator_Metal_titration, Fluka™

Supplier: Honeywell Chemicals

Synonyms: 2,6-Dichlorodimethyl-sulphoxy-fuchsonedicarboxylic acid sodium salt, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid,trisodium salt, Mordant Blue 29, 5-((3-Carboxy-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene)(2,6-dichloro-3-sulphophenyl)methyl)-2-hydroxy-3-methylbenzoic acid, trisodium salt, Chrome Azurol S

72687-25G
72687-25GEA 95.6 GBP
72687-25G
Chromeazurol S (Mordant blue 29) Indicator_Metal_titration, Fluka™
Chromeazurol S (Mordant blue 29)
Formula: C₂₃H₁₆Cl₂O₉S
MW: 605.28 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001615
CAS Number: 1667-99-8
EINECS: 216-787-0

Order Now

Specification Test Results

Identity (IR) complying
Extinction maximum at 424 - 436 nm
Molar absorbancy index Min. 6000


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR