4-Nitroaniline 99,0 HPLC_ASSAY_METHOD Indicator in non-aqueous solvents, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: p-Nitroaniline, pNA

72680-500G
72680-500GEA 324 GBP
72680-500G
4-Nitroaniline 99,0 HPLC_ASSAY_METHOD Indicator in non-aqueous solvents, Fluka™
4-Nitroaniline
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 260 °C
Melting Pt: 146…149 °C
Density: 1.44 g/cm³
Flash Pt: 213 °C
Storage Temperature: Ambient
MDL Number: MFCD00007858
CAS Number: 100-01-6
EINECS: 202-810-1
UN: 1661
ADR: 6.1 II
Merck Index: 12,06681

Order Now

Specification Test Results

Identity (IR) complying
Assay (HPLC) Min. 99.0 %
Melting point 146 - 150 °C
Colour of the solution complying
Clearness of the solution complying
Residue on ignition Max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR