Supelco® Organic Reference Standard, (±)-Citronellal

Supplier: Merck

72638-1ML
72638-1MLEA 60.7 GBP
72638-1ML
Supelco® Organic Reference Standard, (±)-Citronellal
Standards Organic Standards

  • GHS signal: GHS07, GHS09


Caution: P261 - P264 - P272 - P280 - P302 + P352 - P305 + P351 + P338

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65bb41f7d918d5651f06958a62fcb0c5cffd3e6e&vid=65bb41f7d918d5651f06958a62fcb0c5cffd3e6e&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

M-Clarity quality level MQ100
Synonym(s) (±)-3,7-Dimethyl-6-octenal
Environmentally Preferable
Density 0,857 g/ml at 25 °C (lit.)
CAS No. 106-23-0
MDL No. MFCD00038090
Formula (CH₃)₂C=CHCH₂CH₂CH(CH₃)CH₂CHO
Molecular weight 154,25
Boiling point 207 °C (lit.)
Hazard class description H315 - H317 - H319
Flash point 74 °C - Closed cup
Shipping conditions STD
Storage conditions 2…8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR