Counting chambers, Thoma
Supplier: MARIENFELD
0640710 0640711 0640730 0640731
631-0697EA 48.5 GBP
631-0697 720-1898 720-1899 720-1908
Counting chambers, Thoma
Slides Counting Chambers
Designed for the determination of the yeast cell number and IVD applications.
Certifications: IVD directive 98/79/EC.