Mercury(II) sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Mercury sulphate, Mercuric sulphate

7135.1000EA 64.4 GBP
7135.1000 7135.2500
Mercury(II) sulphate, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Mercury (II) sulphate
Formula: HgSO₄
MW: 296.65 g/mol
Melting Pt: 850
Density: 6.47 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011047
CAS Number: 7783-35-9
EINECS: 231-992-5
UN: 1645
ADR: 6.1 II
Merck Index: 13,05914

Order Now

Specification Test Results

Identity (Hg2+) passes test
Molarity (K2Cr2O7) 0.0198 - 0.0202

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR