VWR®, Transport Swabs with Medium
Supplier: VWR Collection
710-0432EA 41.7 GBP
710-0432 710-0433 710-0434 710-0435 710-0436 710-0437 710-0438 710-0439 710-0440 710-0441
VWR®, Transport Swabs with Medium
Swabs Transport Swabs
Collection and transport swabs for bacterial specimens with applicators and transport media.
  • Bacteria will survive for 24 to 48 hours on the applicator and in the tube
  • Tamper-proof seal, which turns white when open
  • Medium risk for transient or short-term use
Suitable for surgical-invasive, short-term applications.
Class IIa - surgical-invasive, short-term application, for taking samples from natural orifices and surgical  wounds.
Certifications: Class IIa.
Packaging: Individually wrapped aluminium pouches under nitrogen ensuring longerMore Product Information