Triethanolamine (trolamine), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor

Synonyms: Tris-(2-hydroxyethyl)amine, 2,2',2''-Nitrilotriethanol

7078.1000EA 73.9 GBP
7078.1000 7078.2500 7078.9025
Triethanolamine (trolamine), BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Triethanolamine (Trolamine)
Formula: C₆H₁₅NO₃
MW: 149.19 g/mol
Boiling Pt: 335.4
Melting Pt: 21.6
Density: 1.13 g/cm³ (20 °C)
Flash Pt: 179 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002855
CAS Number: 102-71-6
EINECS: 203-049-8
Merck Index: 13,09739
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c181d3e80375aec7c34e17766247484e1c3d3257&vid=40c3d3d0c0162ac22208cadb8d5254a7cf0b340a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.0 - 103.0 %
Appearance of solution passes test
Density (g/ml) at 25°C 1.117 - 1.125
Iron (Fe) max. 5 ppm
Heavy Metals (as Pb) max. 10 ppm
Identification passes test
Sulfated Ash max. 0.005 %
Water (H2O) max. 0.2 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR