Hybrid multi-parameter meter, handheld, SL1000 PPA
Supplier: Hach
LPV4439910002 LPV4439920002
705-0941EA 5530 GBP
705-0941 705-0942
Hybrid multi-parameter meter, handheld, SL1000 PPA
Electrochemistry Meters pH and Multi-Parameter Meters
The SL1000 is a portable colorimeter that provides 75% faster testing by using Chemkey® reagents instead of powder pillows or cuvette test methods. No reagent handling is necessary − all chemical and processes are entirely contained inside the Chemkey®. Besides that, the meter can be used in combination with digital IntelliCAL™ electrodes to measure different electrochemical parameters, such as pH, conductivity, ORP, dissolved oxygen, nitrate, fluoride in clean water applications.
  • Ideal for disinfection control in the field or clean water treatment
More Product Information