Hydrogen peroxide 35%, J.T.Baker®

Supplier: Avantor
Danger

7048.1000EA 31.3 GBP
7048.1000 7048.9025
Hydrogen peroxide 35%, J.T.Baker®
Hydrogen peroxide
Formula: H₂O₂
MW: 34.01 g/mol
Boiling Pt: ∼107 °C (1013 hPa)
Melting Pt: < 0 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1 II
Merck Index: 14,04798

Order Now

Specification Test Results

Assay min. 35 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR