Automated solvent extractors according to Randall
Supplier: VELP SCIENTIFIC
F30300242 F30300240
701-0229EA 13570 GBP
701-0229 701-0230
Automated solvent extractors according to Randall
Extractors Automatic Extractor Systems
The SER 148/3 and SER 148/6 can be used to separate a substance or a group of elements (e.g. fat) from solid and semi-solid samples according to the Randall technique (consisting of immersion, washing and solvent recovery). The main field of application is the determination of the content of soluble products such as fats, detergents, plasticisers and pesticides in food, animal feed, detergents, rubber and plastic formulas, pharmaceutical products, soil, and more.
  • Automated process with control of temperature and time of process phases
  • Up to five times
More Product Information