Test strips, drug testing for urine
Supplier: SURESCREEN
AMS BAS BZS CAS MTDS OPS2000 MDMAS TRAMS
700-9544EA 76.2 GBP
700-9544 700-9551 720-0657 700-9564 720-0737 700-9634 720-1305 SURETRAMS
Test strips, drug testing for urine
Drug Tests
Single test strips are an efficient method for testing urine for specific drugs in urine. Simply dip the test strip into a urine sample and read at 3 minutes. Each individual strip is provided in a foil pouch, to ensure the high quality of the product is retained throughout the shelf-life.
  • More than 20 different drug tests available
  • Easy to perform test routine
  • Clear results in 3 minutes
  • Greater than 99% accuracy
  • For detecting recent drug use approx. 1 to 8 days
Recommended markets and use:
On-site screening
Safety critical situations
Random screening
More Product Information