VWR® PerfectBlue Twin L, Dual Gel System

Supplier: VWR Collection
700-1080EA 2420 GBP
700-1080 700-1205
VWR® PerfectBlue Twin L, Dual Gel System
Electrophoresis Systems Vertical Gel Electrophoresis Systems
The largest of the series, the Twin L can accommodate 20×20 cm gels for the simultaneous separation of up to 50 samples making this system ideally suited to either higher throughput or separating lower molecular weight samples. The system actively manages temperature build up with an efficient water cooling system and central aluminium ceramic plates for effective and safe long-term operation.

*Based on an average gel and a tooth at 20 mm depth

Ordering information: Complete system, with or without casting base, includes internal cooling core, SuperSafe cover with attached leads, 4×More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Buffer volume, gel tank ~1250 ml
Sample capacity 10 − 50
Unit W×H×D 300×170×240 mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR