Anti-IgG Sheep Polyclonal Antibody

Supplier: Tonbo Biosciences
70-8078-M002
70-8078-M002EA 127 GBP
70-8078-M002
Anti-IgG Sheep Polyclonal Antibody
Antibodies
Sheep anti-Rabbit IgG (H&L) - Affinity Pure

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and light chain
Host: Sheep
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR