Supelco® BIOshell™ Columns, 160 Å Peptide C18 Fused-Core®

Supplier: Merck
66901-U 66902-U 66903-U 66904-U 66905-U 66906-U 66907-U 66908-U 66909-U 66913-U 66915-U 66918-U 66919-U 66921-U 67001-U 67002-U 67003-U 67004-U 67006-U 67007-U 67008-U 67011-U 67012-U 67013-U 67014-U 67015-U 67016-U 67017-U 67018-U 67093-U 67096-U 67097-U 67099-U 67234-U 67238-U 67239-U 67242-U 67243-U 67257-U 67263-U 67267-U 67274-U 67275-U 67277-U 67281-U 67282-U 67284-U 66922-U 66923-U 67283-U
66901-UEA 543 GBP
66901-U 66902-U 66903-U 66904-U 66905-U 66906-U 66907-U 66908-U 66909-U 66913-U 66915-U 66918-U 66919-U 66921-U 67001-U 67002-U 67003-U 67004-U 67006-U 67007-U 67008-U 67011-U 67012-U 67013-U 67014-U 67015-U 67016-U 67017-U 67018-U 67093-U 67096-U 67097-U 67099-U 67234-U 67238-U 67239-U 67242-U 67243-U 67257-U 67263-U 67267-U 67274-U 67275-U 67277-U 67281-U 67282-U 67284-U 66922-U 66923-U 67283-U
Supelco® BIOshell™ Columns, 160 Å Peptide C18 Fused-Core®
Chromatography Columns
BIOshell™ 160 Å Peptide C18 columns are specifically engineered to provide efficient separation of peptides as well as small proteins.

  • The sterically-protected C18 lighands provide extra stability
  • Higher resolutions and higher peak capacities of peptides at ~50% backpressures of sub-2 µm fully porous paticles (FPP)-packed columns
  • Lower backpressure allows for columns to be used in series to maximize resolutions and peak capacity of complex proteomic samples or tryptic digests
  • Compatible with all UHPLC and LC-MS instrumentation
  • Column can be used at high
More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Application Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides Tryptic Digests; Post-translational Modifications (PTMs); Polypeptides
Carbon load 4,6% 4,6% 4,6%
Endcapped No No No
Max. pressure 600 bar 600 bar 600 bar
pH range 1 - 8 1 - 8 1 - 8
Surface area 60 m²/g 90 m²/g 65 m²/g
Temperature limit 90 °C 90 °C 90 °C
USP code L1 L1 L1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR