Urease (from Canavalia ensiformis)
Supplier: Merck Millipore Calbiochem
666133-10
666133-10EA 196 GBP
666133-10
Urease (from Canavalia ensiformis)
Enzymes
Native urease from Canavalia ensiformis. Catalyzes hydrolysis of urea to carbon dioxide and ammonia. Useful in the determination of urea in body fluids.
  • One unit is defined as the amount of enzyme that will release 1 µmol of ammonia from urea per min at 25 °C.