VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld
Supplier: VWR Collection
665-0494EA 900 GBP
665-0494 665-0495 665-0496
VWR® pHenomenal® MD 8000 H Multi-Parameter Instruments, Handheld
Electrochemistry Meters pH and Multi-Parameter Meters
These digital meters combine six parameters in one instrument: pH, mV, temperature, conductivity, dissolved oxygen and turbidity. IDP technology optimises use and makes measurement easier (no need to recalibrate when changing the meter and no need to adjust the meter’s settings). The measured data is complete with serial number, settings and calibration data.
  • One universal measuring channel
  • Digital sensor recognition: The measured parameter and settings are recognised automatically
  • Suitable for Intelligent Digital Probes (IDP)
  • Suitable for measurements
More Product Information