Dissolved oxygen meters, handheld, Oxi 3205 / 3310
Supplier: WTW
2BA100 2BA101 2BA300 2BA301 2BD350 2BD351 2BD352
664-0525EA 1250 GBP
664-0525 664-0526 664-0533 664-0534 664-0180 664-0140 664-0181
Dissolved oxygen meters, handheld, Oxi 3205 / 3310
Electrochemistry Meters Oxygen Meters
Waterproof handheld meters designed for unlimited outside use.
  • Large display allowing excellent readability, even under bad lighting conditions
  • Robust housing and sealed silicone keypad with protection class IP 66/67
  • Up to 2500 hour continuous operation time with one battery set
  • Large data memory (date, time and ID numbers for measurements)
  • Four user selectable languages available (English, German, French, Spanish)
For challenging measurements in the field and on the plant, e.g. water/wastewater analysis, chemical/pharmaceutical industry, etc.