Conductivity cells, four-electrode, Orion™ DuraProbe™
Supplier: Thermo Orion
013005MD 013010MD 013610MD 013605MD
663-0180EA 507 GBP
663-0180 663-0218 663-0220 663-0189
Conductivity cells, four-electrode, Orion™ DuraProbe™
Electrodes Conductivity Sensors
These durable and extremely accurate conductivity cells can be used under a wide range of conditions. Intended for field or laboratory use.
  • Virtually eliminate fringe field interference errors
  • Rugged epoxy body
  • Graphite sensors
  • Temperature compensated