pH micro electrodes, InLab® Micro / InLab® Nano / InLab® NMR
Supplier: Mettler - Toledo
59904572 51343160 51343161 51343165 51343164
662-1618EA 721 GBP
662-1618 662-2863 662-2866 662-2864 662-1347
pH micro electrodes, InLab® Micro / InLab® Nano / InLab® NMR
Electrodes pH Electrodes
pH electrodes with S7 or BNC connector.
  • ARGENTHAL™ reference system with silver-ion trap (except InLab® Nano / NMR: Ag/AgCl)
  • XEROLYT® EXTRA polymer electrolyte: High signal stability, very short response time and high chemical resistance (InLab® Semi-Micro)
  • InLab® Flex-Micro features a flexible thin shaft (Ø 6 mm): Ideal for measuring pH in test tubes and other small containers
pH range: 0 - 14 for InLab® Micro/Micro Pro/Semi-Micro-L; 0 - 12 for InLab® Semi-Micro
Material: Glass
Ordering information: Supplied with test certificate. Other electrodes available on request.
Order Now
SPECIFICATIONS
Environmentally Preferable