Orion™ Pro Star PI214 pH/Ion Bench Meters
Supplier: Thermo Orion
PSTAR2148 PSTAR2145 PSTAR2140 PSTAR2147 PSTAR2146
662-2517EA 1260 GBP
662-2517 662-2514 662-2513 662-2516 662-2515
Orion™ Pro Star PI214 pH/Ion Bench Meters
Electrochemistry Meters pH and Multi-Parameter Meters
The Thermo Scientific™ Orion™ Pro Star PI214 pH/ion bench meter is designed to provide accurate, reliable electrochemistry testing. Featuring an intuitive interface, enhanced data reporting and robust functionality, the Orion™ Pro Star PI214 meter delivers advanced performance in a modern and simplified package.
  • Large 7" high-resolution colour display
  • Responsive capacitive touchscreen operable with gloves
  • Angled display to help ensure ergonomic viewing
  • Streamlined, easy to operate user interface
  • Vertical meter orientation helps optimise bench space
More Product Information