pH Sensor, Memosens CPS61E
Supplier: Endress+Hauser
CPS61E-10N2/0 CPS61E-11T9/0 CPS61E-1061/0 CPS61E-10R4/0
662-2462EA 664 GBP
662-2462 662-2463 ENDRCPS61E-1061/0 ENDRCPS61E-10R4/0
pH Sensor, Memosens CPS61E
Electrodes pH Electrodes
Memosens 2.0 glass sensor for the food and beverage industry and for bioreactors in biotech industry.
  • Memosens 2.0 offers extended storage of calibration and process data, enabling better trend identification and providing a future-proof basis for predictive maintenance and enhanced IIoT services
  • The sensor's exceptional accuracy and reproducibility help you to keep the pH value in the optimum range for a maximized product yield
  • A special glass membrane and reference system makes the sensor CIP/SIP and autoclaving resistant (up to 140 °C/284 °F) offering
More Product Information