pH combination electrodes, ScienceLine
Supplier: SI Analytics
285100018 285100067 285092329 285092661 285100001 285100034 285100042 285100207 285100215 285100223 285102516 285102524 285102532 285102549 285100059 285101277 285102565 285101211 285101260 285092337 285092591 285092583
662-6104EA 370 GBP
662-6104 662-0517 662-1372 662-2175 662-0512 662-6172 662-6105 662-0513 662-0514 662-0515 662-2182 662-2183 SCOI285102532 662-2184 662-1215 SCOI285101277 SCOI285102565 662-2179 662-0101 662-2172 662-2174 662-2173
pH combination electrodes, ScienceLine
Electrodes pH Electrodes
Heavy-duty combination electrodes for practically all applications in the laboratory and in the field.
  • Maximum measuring accuracy and consistency, with optimum sensor service life
  • Maximum adaptability for all applications, even at high temperatures
  • Enable measurements in deep vessels with a length of up to 600 mm, in MMR tubes with a diameter of 3 mm, and in round bottom flask with NS 14,5 standard taper
* Without cable (available separately).
Electrolyte: KCl 3 mol/l
Shaft material: Glass
Temperature range: −5 to +100 °C (H 61 / H&More Product Information