Combination pH electrode, Orion™ 9102BNWP
Supplier: Thermo Orion
9102BNWP
662-1527EA 220 GBP
662-1527
Combination pH electrode, Orion™ 9102BNWP
Electrodes pH Electrodes
This general purpose pH electrode is ideal for conducting accurate Q.C. and research work.
  • Contains a silver/silver chloride reference, widely used due to accuracy and broad temperature range
  • With waterproof connector
Delivery information: Supplied with 60 ml bottle of Orion electrode fill solution.