Buffer solutions in sachets
Supplier: Mettler - Toledo
51302068 51302080 51302047 51302069 51302070 51302079
662-0540EA 74.5 GBP
662-0540 662-0544 662-0538 662-0541 662-0542 662-0543
Buffer solutions in sachets
Buffers pH Calibration Buffers
Solutions in sachets ensure optimum precision.
  • Advantages
  • No danger of contamination
The plastic-coated sachet provides a perfect liquid seal and good protection from light. The solutions should only be used once to avoid any danger of contamination. They also satisfy the requirements of the GLP and ISO standards. They have been adjusted using a pH-metre that has been calibrated with a buffer associated with NIST standards.
Delivery information: The buffers in sachets are delivered with a certificate of conformity.
Order Now
SPECIFICATIONS
Environmentally Preferable