Combined ion selective electrodes, 500 series
Supplier: WTW
106395
662-0381EA 1160 GBP
662-0381
Combined ion selective electrodes, 500 series
Electrodes ISE Electrodes
These 500 series half cells require a separate reference electrode type R 503 (an exception for this is the gas-sensitive NH 500/2 with integrated reference electrode).
  • Ideal for challenging ISE applications
Ordering information: Reference electrodes, ionic strength adapters and standard solutions are not included. Please contact Avantor for further information.