n-Heptane 99%, CHROMASOLV™ Plus HPLC, Riedel-de Haën™

Supplier: Honeywell Chemicals
Danger

Synonyms: high purity solvents

650536-1L
650536-1LEA 117 GBP
650536-1L
n-Heptane 99%, CHROMASOLV™ Plus HPLC, Riedel-de Haën™
n-Heptane

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities

Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.2…98.4 °C (1.000 hPa)
Melting Pt: –90.5 °C
Density: 0.69 g/cm³ (15 °C)
Flash Pt: 6.7 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Autoignition Temperature 433 °F
Purity (GC) Min. 98.5 %
IR spectrum complying
Refractive index (n 20/D) 1.3850 - 1.3890
Non-volatile matter Max. 0.0001 %
Water (Karl Fischer) Max. 0.01 %
Absorbance at 400 nm Max. 0.005
Absorbance at 300 nm Max. 0.005
Absorbance at 250 nm Max. 0.01
Absorbance at 225 nm Max. 0.10
Absorbance at 200 nm Max. 1.0
Fluorescence (chinin) at 365 nm Max. 1.0 ppb
GC (as heptachlor epoxide) Max. 2 µg/l
Vapor Density 3.5 (vs air)
Vapor Pressure 40 mmHg ( 20 °C)
Explosion Limit 7 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR