Handheld refractometers with special scales, MASTER series
Supplier: Atago
2612 2481 2483 2930 2931 2932 2491 2493 2691 2471 2473
635-0709EA 166 GBP
635-0709 635-0497 635-0705 635-0711 635-0498 635-0712 635-0706 635-0707 635-0710 635-0703 635-0704
Handheld refractometers with special scales, MASTER series
Refractometers Handheld Refractometers
The MASTER series of handheld refractometers provides a range of easy to use instruments for a wide variety of applications.
  • MASTER-M: Standard model, clear scale, only two drops required
  • MASTER-T: Automated temperature compensation from 10 to 30 °C
  • MASTER-α: Automated temperature compensation (IP 65 except eyepiece), dust-tight and protected against water jets
  • MASTER-P: Designed with a plastic body for resistance to salty and acidic samples
The design has an ergonomic grip and a prism housing that allows for the sample to cover the prism withoutMore Product Information