VWR® UV-1600PC, UV/Visible Spectrophotometer
Supplier: VWR Collection
634-6001EA 2930 GBP
634-6001
VWR® UV-1600PC, UV/Visible Spectrophotometer
Spectrophotometers and Photometers UV/Visible Spectrophotometers
The UV-1600PC is designed for routine daily quality control requirements, in-process control and teaching laboratories.
  • Can save up to a total of 200 results and 200 standard curves
  • Download data to PC via USB interface
  • Optional accessories including an automatic eight-cell changer
The application software also offers all the main routine measurement modes.
Delivery information: Supplied with four optical glass cells, two quartz cells, four-position cell holder, power cable with UK and EU plug, dust cover and application software for PC control.