Magnesium sulphate, anhydrous 98,0 Complexometric, powder_extra_fine, Puriss. p.a. Drying_agent, Fluka™

Supplier: Honeywell Chemicals

Synonyms: MgSO4, Magnesium sulphate anhydrous

63136-250G 63136-1KG 63136-25KG
63136-250GEA 87.7 GBP
63136-250G 63136-1KG HONC63136-25KG
Magnesium sulphate, anhydrous 98,0 Complexometric, powder_extra_fine, Puriss. p.a. Drying_agent, Fluka™
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1.130 °C (dec.)
Density: 2.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Form powder (very fine)
Assay 98.0 - 102.0 %
Loss on ignition (600°C) Max. 2 %
Aluminium (Al) Max. 20 ppm
Arsenic (As) Max. 0.1 ppm
Barium (Ba) Max. 5 ppm
Bismuth (Bi) Max. 5 ppm
Calcium (Ca) Max. 400 ppm
Cadmium (Cd) Max. 5 ppm
Cobalt (Co) Max. 5 ppm
Chromium (Cr) Max. 5 ppm
Copper (Cu) Max. 5 ppm
Iron (Fe) Max. 10 ppm
Potassium (K) Max. 200 ppm
Lithium (Li) Max. 5 ppm
Manganese (Mn) Max. 20 ppm
Molybdenum (Mo) Max. 5 ppm
Sodium (Na) Max. 100 ppm
Nickel (Ni) Max. 5 ppm
Lead (Pb) Max. 5 ppm
Strontium (Sr) Max. 5 ppm
Zinc (Zn) Max. 5 ppm
Chloride (Cl) Max. 50 ppm
Nitrate (NO₃) Max. 40 ppm
Ammonium (NH₄) Max. 20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR