Linoleic acid, Supelco®

Supplier: Merck

Synonyms: (9Z,12Z)-Octadeca-9,12-dienoic acid, cis,cis-9,12-Octadecadienoic acid, 9Z,12Z-Octadecadienoic acid

MQ100

62230-25ML-F 62230-5ML-F
62230-25ML-FEA 251 GBP
62230-25ML-F 62230-5ML-F
Linoleic acid, Supelco®
Linoleic acid
Formula: C₁₈H₃₂O₂
MW: 280.45 g/mol
Boiling Pt: 360
Melting Pt: –5 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064241
CAS Number: 60-33-3
EINECS: 200-470-9
Merck Index: 13,05527
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e249c7776ab623cf99dd56e541ef4d8bd0b5c7e&vid=d706be2418dfecf2812ea64dd7f2128e2448bc49&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR