(S)-(-)-Limonene, Supelco®

Supplier: Merck
Danger

Synonyms: (-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene

MQ100

62128-1ML 62128-5ML
62128-1MLEA 41.7 GBP
62128-1ML 62128-5ML
(S)-(-)-Limonene, Supelco®
(S)-(-)-Limonene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 175
Melting Pt: –74 °C
Density: 0.84 g/cm³ (20 °C)
Flash Pt: 43
Storage Temperature: Ambient
MDL Number: MFCD00001558
CAS Number: 5989-54-8
EINECS: 227-815-6
UN: 2052
ADR: 3 III
Merck Index: 13,05515
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=925593852eaaa99e409db79716af7534bbb3f96e&vid=5d7cea0c73faf38c4a26b1dd60ffc962e4f141f9&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR