Methyl ethyl ketone 99, ChromAR® LC, UV, Macron Fine Chemicals™

Supplier: Avantor
Danger

Synonyms: EK 500 Additive, Ethyl methyl ketone, 2-Butanone, Butanone, MEK

M.O.C. Category N

6206-25EA 0 GBP
6206-25
Methyl ethyl ketone 99, ChromAR® LC, UV, Macron Fine Chemicals™
Methyl ethyl ketone
Formula: C₂H₅COCH₃
MW: 72.11 g/mol
Boiling Pt: 79.6 °C (1013 hPa)
Melting Pt: –86 °C
Density: 0.8054 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3 II
REACH: 01-2119457290-43
Merck Index: 13,06097

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
ACS - Assay (CH₃COCH₂CH₃) (by GC) ≥99.0 %
ACS - Color ≤15
ACS - Residue after Evaporation ≤5 ppm
ACS - Water (H₂O)(by Karl Fischer titrn) ≤0.08 %
Optical Absorbance (1-cm path vs water) - 329 nm ≤1.00
Optical Absorbance (1-cm path vs water) - 335 nm ≤0.50
Optical Absorbance (1-cm path vs water) - 344 nm ≤0.05
Optical Absorbance (1-cm path vs water) - 350 - 400 nm ≤0.01
Titrable Acid (meq/g) ≤0.0005
Filtered through a 0.2 micron filter.
Appearance (clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR