Digital barometer, 511
Supplier: Testo
0560.0511
620-3303EA 173 GBP
620-3303
Digital barometer, 511
Barometers
Pocket absolute pressure meter. Very handy, small and easy to use, this barometer permits measurements of barometric air pressure by entering on-site height above sea level and barometric elevation measurement between two points. Backlit display completes the instrument.
  • Automatic off function
  • Eight switchable pressure units: hPa, mbar, Pa, mmH₂O, mmHg, inH₂O, inHg, psi
  • Barometric altitude measurement
Delivery information: Supplied with protective cap, wrist strap, belt holder and two alkaline AAA batteries for about 500 hours of use without display illumination.