Digital thermo-hygrometers, TFH 610
Supplier: EBRO
1340-5610
620-1393EA 292 GBP
620-1393
Digital thermo-hygrometers, TFH 610
Hygrometers Digital Thermo-Hygrometers
These robust and impact resistant thermo-hygrometers provide high accuracy measurements for monitoring storage checks.
  • °C/°F switchable
  • Capacitive sensor humidity
  • Two measurement channels
  • Automatic deactivation
Thermistor sensor.
Delivery information: Supplied with a sensor, a six-point factory calibration certificate, a Lithium three-volt battery for up to five years of life, with a charge indicator.