Ubbelohde viscometers, uncalibrated
Supplier: SI Analytics
285400304 285400329 285400337 285400345 285400353 285400361 285400378 285400386
616-3347EA 395 GBP
616-3347 616-3349 616-3350 616-3351 616-3352 616-3353 616-3354 616-3355
Ubbelohde viscometers, uncalibrated
Viscometers Capillary Viscometers
Viscometers with suspended ball level for determination of relative viscosity of liquids with Newtonian flow behaviour.
Recommended minimum flow-through time is 200 seconds.
Not calibrated to measure relative viscosity.
Certifications: DIN 51 562 Part 1, ISO/DIN 3105 (BS-IP-SL). Without calibration certificate.