Multi-channel pipettes, mechanical, variable volume, Tacta
Supplier: Sartorius
LH-729120 LH-729130 LH-729140 LH-729220 LH-729230 LH-729240
613-5486EA 878 GBP
613-5486 613-5487 613-5488 613-5489 613-5490 613-5491
Multi-channel pipettes, mechanical, variable volume, Tacta
Pipettes Micropipettes
This mechanical pipette with large display has been designed with comfort in mind. Tacta makes pipetting effortless and safe, while producing accurate and reliable results. The pipette is made from carefully selected materials, with each component designed to meet the highest standards.
  • Comfortable to hold due to the ergonomically designed handle
  • Low pipetting and tip ejection forces that reduce the risk of Work Related Upper Limb Disorder (WRULD)
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and smooth
More Product Information