Single-channel pipettes, mechanical, fixed / variable volume, Proline® Plus
Supplier: Sartorius
728010 728020 728030 728050 728060 728070 728080 728090 728515 728520 728530 728535 728545 728550 728560 728565 728567 728570 728580 728590 728040
613-5160EA 247 GBP
613-5160 613-5161 613-5162 613-5163 613-5164 613-5165 613-5166 613-5167 613-5168 613-5169 613-5170 613-5171 613-5172 613-5173 613-5174 613-5175 613-5176 613-5177 613-5178 613-5179 613-1753
Single-channel pipettes, mechanical, fixed / variable volume, Proline® Plus
Pipettes Micropipettes
High quality pipettes with new, innovative design and technology, combining safety, ergonomics, efficiency in pipetting, with easy calibration and maintenance. They include built-in replaceable tip cone filters, which prevent pipette contamination and damage.
  • Good ergonomics - handle design suitable for both left- and right-handed users, clear and easy to read display with big numbers and small increments, light and easy tip loading and ejection
  • Fit the most commonly used lab tips thanks to slim tip ejector collar
  • Plunger technology results in the low
More Product Information