Single channel pipettes, mechanical, fixed / variable volume, Proline®
Supplier: Sartorius
720015 720080 720025 720050 720070 720060 720110 722001 722004 722010 722015 722020 722025 722030 722035 722040 722045 722050 722055 720010
613-4045EA 241 GBP
613-4045 613-4053 613-4047 613-4050 613-4052 613-4051 613-4054 613-4077 613-4078 613-4079 613-4080 613-4081 613-4082 613-4083 613-4084 613-4085 613-4086 613-4087 613-4088 613-4046
Single channel pipettes, mechanical, fixed / variable volume, Proline®
Pipettes Micropipettes
Ergonomic pipettes with fast, easy operation. Selected models offer changeable tip cone filters to prevent liquids and aerosols entering the pipette.
  • Easy calibration and maintenance, small number of parts
  • Smooth plunger action and simple in-lab recalibration and service
  • Volume setting by a simple click-stop
  • Ergonomic finger support, which reduces the risk of RSI
Two types of filters are offered: Safe-Cone standard filter and Safe-Cone Plus filter. It is recommended to use the Standard filter for general applications and the Plus filter for moreMore Product Information