Filter tips, sterile, ep Dualfilter T.I.P.S®
Supplier: EPPENDORF
0030077.555 0030077.547RCH
613-3685EA 0 GBP
613-3685 EPPE0030077.547RCH
Filter tips, sterile, ep Dualfilter T.I.P.S®
Pipette Tips
Two-phase filter for ultimate aerosol protection: The white layer which faces the sample retains drops, splashes and aerosols; the blue layer above it forms a highly effective double-barrier, that reliably binds even biomolecules.
  • Characterised by defined flow dynamics, low wettability and high thermal stability
  • Certified pyrogen-free and PCR clean (free from human DNA, DNase, RNase and PCR-inhibiting substances)