Gastight® 1000 series syringes
Supplier: HAMILTON BONADUZ
201200 201250 201300 201350 201400 201700 81301 81316 81317 81320 81322 81323 81330 81343 81401 81414 81416 81417 81420 81443 81501 81514 81516 81517 81520 81543 81601 81616 81617 81620 81643 82525 81427 81314 81630 7651-01 7649-01 7650-01 7652-01 81341 81327 81430 81527 81530 81627 85020 86020 201812 85021 85025 85027 82520 82527 81365 81560 81570 81610 81660 CAL81520 CAL81417 CAL82520 CAL81327 CAL81527 CAL81316 CAL81317 CAL81416 CAL7649-01 CAL81517 CAL81343 CAL81301 CAL81420 CAL81365 CAL81430 CAL81627 81441 CAL81401 CAL81516 CAL81616 CAL81620 208645 CAL81330 CAL81443 CAL81320 CAL85021 CAL86020 CAL7650-01 CAL7651-01 CAL81314 CAL81543 CAL81501
613-1605EA 124 GBP
613-1605 613-0108 548-2264 613-1607 548-2265 613-1132 549-1189 613-1300 549-1190 613-1321 549-0231 613-1331 549-0369 549-1191 549-1192 548-1967 549-0370 613-1302 613-1322 549-1193 549-1194 548-2318 548-2319 613-1303 613-1323 549-1195 549-1196 613-1304 549-0232 613-1324 548-2324 613-1325 548-1968 548-1966 549-1267 548-2279 548-2429 548-2278 548-2430 548-2316 548-2315 549-1265 548-2320 549-1266 548-2323 613-1326 613-1327 548-1959 548-2326 548-2327 548-1972 548-1971 548-2325 613-1166 548-2321 548-2322 548-1969 548-1970 548-2352 548-2349 548-2448 548-2436 548-2444 548-2346 548-2347 548-2440 548-2431 548-2351 548-2437 548-2345 548-2350 548-2438 548-2441 548-2447 548-2317 548-2439 548-2443 548-2446 548-2353 548-2271 548-2348 548-2442 548-2435 548-2449 548-2450 548-2432 548-2433 548-2434 548-2445 HAMICAL81501
Gastight® 1000 series syringes
Syringes Syringes for Chromatography
The 1000 series is a mid volume Gastight® syringe. Gastight® syringes are ideal for dispensing both liquids and gases. They have a precision machined PTFE plunger tip which creates a leak-free seal. With the tight fit, the tip essentially wipes the interior of the syringe barrel free of sample. This feature is particularly useful with heterogeneous samples as it reduces the chance that a deposit will occur and cause the plunger to freeze.
  • Ideal for dispensing both liquids and gases in volumes from 100 µl up to 100 ml
Removable needle (RN): The designMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable