Measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Supplier: Brand
32162 32164 32105 32120 32128 32138 32148 32154 32108
612-5022EA 102 GBP
612-5022 612-4118 612-5015 612-5017 612-5018 612-5019 612-5020 612-5021 612-5016
Measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Cylinders Measuring Cylinders
Borosilicate glass 3.3, tall form, class A, DE-M marking.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, with spout
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
DIN EN ISO 4788
Certifications: DIN EN ISO 4788.
Ordering information: Supplied with a batch certificate. Measuring cylinders are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.
Order Now
SPECIFICATIONS
Environmentally Preferable