Bulb pipettes (one mark), BLAUBRAND®, Class AS, USP
Supplier: Brand
30607 30619 30618 30611 30612 30610 30609 30608 30617 30616 30613 30603 30606 30605 30602 30601 30615 30614
612-4789EA 36 GBP
612-4789 612-4801 612-4800 612-4793 612-4794 612-4792 612-4791 612-4790 612-4799 612-4798 612-4795 612-4786 612-4788 612-4787 612-4785 612-4760 612-4797 612-4796
Bulb pipettes (one mark), BLAUBRAND®, Class AS, USP
Pipettes Bulb Pipettes
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • Ring mark and inscriptions in blue enamel
  • Waiting time 5 seconds
  • Incl. USP batch certificate, also available with USP individual certificate on request
Certifications: DIN EN ISO 648; DE-M marking.