Measuring cylinders, tall form, class B, PMP
Supplier: Brand
34720 34728 34748 34762 34908 34928 34948 34954 34962
612-4326EA 9.8 GBP
612-4326 612-4327 612-4329 612-4331 612-4631 612-4633 612-4635 612-4636 612-4637
Measuring cylinders, tall form, class B, PMP
Cylinders Measuring Cylinders
PMP, transparent, tall form, class B.
  • Calibrated to contain (TC, In)
  • With blue printed scale or embossed scale (embossed scale recommended for autoclaving)
  • Exposure to temperatures up to 121 °C will not cause permanent exceeding of tolerance limits
  • Cleaning temperatures below 60 °C is recommended to preserve marks and inscriptions
DIN 12 681 / ISO 6706
Certifications: DIN 12 681 / ISO 6706