VWR®, Bottle-Top Dispensers
Supplier: VWR Collection
612-4750EA 311 GBP
612-4750 612-4751 612-3669 612-3670 612-3671 612-3672
VWR®, Bottle-Top Dispensers
Bottle Top Dispensers
Variable volume, autoclavable.
  • Valve system with steel spring
  • PP with precision glass cylinder
  • Quick and easy to use volume setting system
  • Additional rotating fine volume adjustment for right or left handed use
  • Adapter allows rotation of the dispenser on bottles to the desired position
These standard PP units, with glass cylinder, are suitable for aqueous liquids, dilute acids and alkalis - not for use with solvents and hydrofluoric acid which attack PP (for bottles with GL 32 thread). The glass cylinder and PTFE-encased glass piston, with PP body,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable