Potassium chloride, Supelco®

Supplier: Merck

Synonyms: Potassiumchloride

M-Clarity™ quality level = MQ100

60136-250ML
60136-250MLEA 85 GBP
60136-250ML
Potassium chloride, Supelco®
Potassium chloride
Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Density: 1.98 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=276efa8c12a5b438884022d6a6694bce46b2724f&vid=16f79dd307ab37e110215bfc332384f50cd5cfa2&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR