Discovery® C8 HPLC Columns
Supplier: Merck
59354-U 59589-U 577501-U 59588-U 569426-U 569427-U 59352-U 59353-U 59353-U21
59354-UEA 786 GBP
59354-U 59589-U 577501-U 59588-U 569426-U 569427-U 59352-U 59353-U 59353-U21
Discovery® C8 HPLC Columns
Chromatography Columns
The Discovery® C8 columns provide the best suitable selectivity for many applications.
  • Less hydrophobic reversed-phase selectivity
  • Retention with excellent peak shape for all compounds
  • Very high stability and no-bleeding properties for LC-MS application
  • Impurity: <10 ppm metals
  • Surface coverage: 3,4 µmol/m²
Designed to meet the exacting requirements of pharmaceutical analysis and purification. Discovery® columns are also ideal for all application segments requiring reversed phase HPLC.
Ordering information: Column test certificate available inside the box.