Septum adaptor, b.safe
Supplier: Bohlender
M508-01
590-0182EA 41.1 GBP
590-0182
Septum adaptor, b.safe
Fittings and Connectors
Threaded adaptor to connect a septum cap with a capillary connector of b.safe caps.
  • Easy sampling
  • Without contamination
  • Without evaporation
Easy sampling on the capillary connectors of b.safe caps and waste caps without contamination or evaporation of solvents.